Learn More
Epredia™ Mounting Medium
Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.
Brand: Epredia™ 4111
97.75 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Rapid drying
- Toluene-based
- Optically clear
Specifications
1 pt. | |
2/Cs. | |
Mounting Medium |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 3
- Respiratory sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Germ cell mutagenicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : toluene, acrylic resin