Learn More
RO-9187, MedChemExpress
MedChemExpress RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.
Brand: Medchem Express HY-10870
Description
RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.Specifications
RO-9187 | |
Light Yellow | |
C9H12N6O5 | |
DMSO : 100 mg/mL (351.83 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (25.12 mM; Need ultrasonic) | |
284.23 | |
98.0% | |
Solid |
876708-03-1 | |
Research | |
50 mg | |
O=C1N=C(N)C=CN1[C@H]2[C@@H](O)[C@H](O)[C@](CO)(N=[N+]=[N-])O2 | |
284.23 | |
Research | |
COVID-19-anti-virus |