Learn More
Quickfit™ Plain Side-Arms
Design gives excellent flow control and allows use with fluids and gases having temperatures between -20°C and 200°C.
Brand:  Quickfit GP6RA/7
 View more versions of this product
Product Code. 12957306
  • 336.00 € / Pack of 5
Estimated Shipment: 06-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Rotary design for fine single drop control invaluable on burettes
  • Borosilicate glass with PTFE key for excellent chemical resistance
  • No lubrication required - no seizure and no contamination
  • Temperature range -20°C to 200°C
  • Vacuum capability to 10-4Torr
  • Increased barrel diameter for excellent drainage
  • Locking ring to prevent accidental removal
  • Ergonomic key for good grip and easy operation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plain Side-arm
10 mm
Borosilicate Glass, PTFE (Key)
GP Rotaflo™ stopcock