Learn More
Quickfit™ Plain Side-Arms
Design gives excellent flow control and allows use with fluids and gases having temperatures between -20°C and 200°C.
215.00 € - 1015.00 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
12485266
|
Quickfit
GP3RA/5 |
8 mm |
215.00 €
Pack of 5 |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||||
12957306
|
Quickfit
GP6RA/7 |
10 mm |
336.00 €
Pack of 5 |
Estimated Shipment: 05-03-2025
Log in to see stock. |
|||||
15577910
|
Quickfit
GP10RA/10 |
13 mm |
1015.00 €
Pack of 10 |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
Description
- Rotary design for fine single drop control invaluable on burettes
- Borosilicate glass with PTFE key for excellent chemical resistance
- No lubrication required - no seizure and no contamination
- Temperature range -20°C to 200°C
- Vacuum capability to 10-4Torr
- Increased barrel diameter for excellent drainage
- Locking ring to prevent accidental removal
- Ergonomic key for good grip and easy operation
Specifications
Plain Side-arm |