Learn More
Hirschmann Instruments™ rotarus™ Volume 50 Peristaltic Pump
Facilitates small delivery volumes and high viscosities for precise pumping in the laboratory. Hirschmann Instruments™ rotarus™ Volume 50 Peristaltic Pump offers superior continuous delivery in a dispensing pump across a broad range of applications.
Brand:  Hirschmann Instruments™ 9501362
Product Code. 15395423
  • 5475.00 € / Each
Explore more special offers
Description
Description
  • Changeable pump heads, single and multichannel design
  • Rapid tool-free changing of pump heads
  • Automatic pump head and hose detection with RFID technology
  • Control panel located directly on pump housing
  • TFT touchscreen with user-friendly menu
  • Calibratable
  • Method memory
  • Operation with 24 V DC
  • USB and RS232 interface for PC link using interface (available as accessory)
  • Pump heads from other manufacturers can be fitted without difficulty using an adapter
  • Comprehensive accessory range
Specifications
Specifications
1 to 12 (Depending on the pump head type used)
205 x 100 x 220 mm
100/240 VAC
Stepper motor
7.3 kg
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Hirschmann Instruments™ rotarus™ Volume 50 Peristaltic Pump >