Learn More
Fisherbrand™ Rotary Vane Vacuum Pump
Designed for laboratory applications requiring deep vacuum. Fisherbrand™ Rotary Vane Vacuum Pump is a two-stage direct driven rotary vane pumps that provide a high water vapor tolerance.
Brand: Fisherbrand™ 302105
This item is not returnable.
View return policy
Description
- Two-stage direct driven rotary vane pump
- Designed for wide range of physical and chemical laboratory applications and continuous running
- Low weight, small footprint and high pumping speed, increased chemical resistance are advantageous features in research and routine processes
- Rugged reliable design, combined with low noise level and ease of use
- Innovative gas ballast valve prevents moisture condensing and high tolerance of water and solvent vapors
- Intake line is vacuum sealed by and anti suckback valve to protect your application from undesired venting and oil back flow after pump is turned off
- Vertical oil sight glass allows easy monitoring of oil level and condition
Specifications
1 x 10-2 mbar ultimate vacuum | |
30 x 12 x 15 cm (11.8 x 4.7 x 5.9 in.) | |
17.6 lb. | |
30/36 L/min. | |
50/60 Hz | |
30 L/min. | |
100/230 V |
230 V 50/60 Hz motor with Schuko and UK power leads | |
90 w | |
8 kg | |
Labovac 10 | |
Rotary Vane Vacuum Pump | |
High Vacuum Distillation |