Learn More
Scientific Industries Genie Roto-Shake Rotator/Rocker
The Genie Roto-Shake Rotator/Rocker with multi-action platform performs rocking or rotating motions. Magnetic stainless steel strips and spring-clip mounting plates offer a variety of configurations for different applications. Wide ambient range is ideal for use in cold rooms and incubators.
Brand: Scientific Industries 51603-05
1579.86 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- Now you can provide a host of shaking options for your applications with the addition of this all-in-one shaker unit.
- It easily changes from rocking to rotating, and the magnetic platform may be removed and replaced with a 3-D orbital shaking platform.
- A simple mechanical adjustment changes the rocker/rotator function.
- The moving component of this shaker is a two-sided 8 in. W x 13.5 in. L stainless steel platform.
- This accommodates a wide range of attachments that mount in a variety of configurations for different applications.
- Use magnetic SS strips to mount membrane-based blot assays/washings directly on platform.
- The non-skid metal rocker platform attachment for trays, and the autoclavable spring-clip mounting plates for three different size tubes fasten magnetically to the platform.
- The stainless steel platform is easy to clean and decontaminate.
- Because shaker maintains set speeds between 0 and 38°C, it is suitable for use in cold rooms and incubators.
- For greater capacity, order expansion kits separately.
Specifications
±10° | |
±10° | |
240 V | |
38°C | |
100.4°F | |
342 x 203.2 mm | |
342 mm | |
8 in. | |
203.2 mm | |
4.5 kg |
4.5 kg | |
5 to 35 rpm | |
240 V | |
9.9 lb. | |
50/60 Hz | |
13.5 in. | |
19.9 in. | |
505 mm | |
9.9 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.