Learn More
Thermo Scientific™ Rotor, 4-Place Swinging Bucket, for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ Centrifuge
For use with Legend Mach 1.6 and 1.6R Centrifuges
Brand:  Thermo Scientific™ 75002000
2358.23 EUR valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Product Code. 10502173
  • 2482.35 € / Each
Explore more special offers
Description
Description
  • Rotor body only; buckets and biocontainment lids sold separately
  • Choose rectangular or round buckets
  • Adapters sold separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor
4,618 x g
Legend Mach 1.6 Series
4 x 400 mL
4,700 rpm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Rotor, 4-Place Swinging Bucket, for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ Centrifuge > Centrifuge Acc,rotor 4 X 400mL