missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors

Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.

Brand:  Thermo Scientific™ 75004083

69.24 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 15374194

  • 77.69 € / 1 unit

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 x 1.5/2.0mL Rotor
Fixed Angle Rotor
9,800 x g
mySPIN 12 Mini Centrifuge
Reinforced Glass Fiber
12 x 1.5/2 mL
12,500 rpm
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors > 12 x 1.5/2.0 mL tube rotor, for mySPIN 12 Mini centrifuge

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.