Learn More
Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation
Ideal for routine laboratory analysis and procedures. Lenz Laborglasinstrumente™ Test Tubes with NS Joint, without Graduation are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05700107
This item is not returnable.
View return policy
Description
- With round bottom and NS socket
- Available without stopper, with PE stopper, or with hexagonal hollow glass stopper
- Corrosion-free and resistant to thermal shock
Specifications
Glass Tube | |
Neck Size: 14/23; Material: Polyethylene; Without Stopper |