Learn More
Eppendorf™ Round Buckets for S-4x400 Rotor
These Round Buckets for S-4x400 Rotor are suitable swing-buckets for the S-4x400 rotor that feature a round shape and a 400 mL capacity.
979.90 € - 1854.39 €
Specifications
For Use With (Equipment) Rotor S-4x400
Capacity 400 mL
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15853212
View Documents
Eppendorf™
5895183000
2 Pk.
979.90 €
Pack of 2
15843212
View Documents
Eppendorf™
5895182003
4 Pk.
1854.39 €
Pack of 4
Description
Description
  • Suitable for S-4x400 rotor
  • Round shape
  • Swing-out bucket
  • Variable places depending on adapter used
Specifications
Specifications
Rotor S-4x400
400 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Round Buckets for S-4x400 Rotor