Learn More
Eppendorf™ Round Buckets for S-4x400 Rotor
These Round Buckets for S-4x400 Rotor are suitable swing-buckets for the S-4x400 rotor that feature a round shape and a 400 mL capacity.
Brand: Eppendorf™ 5895182003
Description
- Suitable for S-4x400 rotor
- Round shape
- Swing-out bucket
- Variable places depending on adapter used
Specifications
Rotor S-4x400 | |
400 mL | |
4 Pk. |
Product Suggestions
Customers who viewed this item also viewed.