Learn More
Abeba™ Rubber 5200 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5200/38
55.64 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 15306691
  • 72.50 € / 1 pair
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB SRC
  • Smooth leather white
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Specifications
CE EN ISO 20347:2012 OB SRC
Leather
Slip-resistant
5200
White
38
Rubber