missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rubber Pipette Bulbs for Bulb or Cylindrical Pipettes

Excellent for all media that do not attack rubber. Fisherbrand™ Pipette Bulbs come with three glassball valves, adjustable by finger pressure.

Brand:  Fisherbrand™ 4709922

 View more versions of this product

Product Code. 11880950

  • 6.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in a range of colors
  • With ca. 60mL volume suitable for pipettes from 2 to 50mL
  • Pipette media must not enter pipette filler; it may cause valve malfunction
Specifications

Specifications

Green
Pipette Filler
Can be used on bulb or cylindrical pipettes from 2mL to 50mL
Rubber
60 mL
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.