missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 2% on 3.18mm (0.125in) alumina pellets, Thermo Scientific Chemicals

122.61 € - 2127.59 €

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11415350
SDS View Documents
Thermo Scientific Alfa Aesar
044575.14
25 g
122.61 €
25g
Please to purchase this item. Need a web account? Register with us today!
11425350
SDS View Documents
Thermo Scientific Alfa Aesar
044575.22
100 g
460.29 €
100g
Please to purchase this item. Need a web account? Register with us today!
11435350
SDS View Documents
Thermo Scientific Alfa Aesar
044575.36
500 g
2127.59 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-18-8
101.07
KJTLSVCANCCWHF-UHFFFAOYSA-N
23950
ruthenium
Ru
MFCD00011207 MFCD03458417
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
CHEBI:30682
[Ru]

Specifications

7440-18-8
MFCD00011207 MFCD03458417
KJTLSVCANCCWHF-UHFFFAOYSA-N
ruthenium
23950
Odorless
Ru
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
[Ru]
101.07
CHEBI:30682
Ruthenium, 2% on alumina pellets, 3.18mm (0.125 in.)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-127-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.