missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals Product Code.: 15750829

Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals

Product Code. 15750829
50 g, Glass bottle
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 15750829

Brand: Thermo Scientific Acros 197210500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-08-8
Molecular Formula Cl3Ru
Molecular Weight (g/mol) 207.43

Specifications

Melting Point >500.0°C
Color Black
Physical Form Crystals or Powder
Quantity 50 g
Assay Percent Range 48 to 50% (Ru)
Linear Formula RuCl3
Fieser 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292
Merck Index 15,8436
Solubility Information Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol
Formula Weight 207.43
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ruthenium(III) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464261 Certificate TypeCertificate of Analysis Date16/07/2024 Catalog Number197210100,197210020,197210500
Lot NumberA0462555 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number197210100,197210020,197210500
Lot NumberA0458807 Certificate TypeCertificate of Analysis Date15/12/2023 Catalog Number197210100,197210020,197210500
Lot NumberA0442229 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number197210100,197210020,197210500
Lot NumberA0443923 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number197210100,197210020,197210500
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.