Learn More
Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals
154.77 € - 568.83 €
Chemical Identifiers
CAS | 18902-42-6 |
---|---|
Molecular Formula | Cl3NORu |
Molecular Weight (g/mol) | 237.43 |
MDL Number | MFCD00049765 |
InChI Key | AUWSZCCJRSCQEP-UHFFFAOYSA-K |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327198
|
Thermo Scientific Alfa Aesar
016622.03 |
1 g |
154.77 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11317198
|
Thermo Scientific Alfa Aesar
016622.06 |
5 g |
568.83 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ruthenium nitrosyl chloride is an important ruthenium compounds, which is used in synthesis of more complex ruthenium compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18902-42-6 | |
237.43 | |
AUWSZCCJRSCQEP-UHFFFAOYSA-K |
Cl3NORu | |
MFCD00049765 |
Specifications
18902-42-6 | |
Cl3NORu | |
Insoluble in water. | |
237.43 | |
Ru 42.1% min. | |
Ruthenium(III) nitrosylchloride |
Crystalline | |
MFCD00049765 | |
AUWSZCCJRSCQEP-UHFFFAOYSA-K | |
237.44 | |
Hygroscopic |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 242-651-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only