missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-1-Phenyl-1,3-propanediol, 98%, Thermo Scientific Chemicals
50.60 € - 578.00 €
Chemical Identifiers
CAS | 96854-34-1 |
---|---|
Molecular Formula | C9H12O2 |
Molecular Weight (g/mol) | 152.19 |
MDL Number | MFCD00221510 |
InChI Key | RRVFYOSEKOTFOG-VIFPVBQESA-N |
Synonym | 1s-1-phenylpropane-1,3-diol, s-1-phenyl-1,3-propanediol, s-1-phenylpropane-1,3-diol, 1s-phenyl-1,3-propanediol, s-1-phenyl-propane-1,3-diol, s, ?-1-phenyl-1,3-propanediol, 1,3-propanediol,1-phenyl-, 1s, 1,3-propanediol, 1-phenyl-, 1s, s---1-phenyl-1,3-propanediol, puriss sum of enantiomers, gc |
PubChem CID | 6950537 |
IUPAC Name | (1S)-1-phenylpropane-1,3-diol |
SMILES | C1=CC=C(C=C1)C(CCO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10740755
|
Thermo Scientific Acros
417762500 |
250 mg | Glass bottle |
50.60 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10045810
|
Thermo Scientific Acros
417760010 |
1 g | Glass bottle |
150.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11964651
|
Thermo Scientific Acros
417760050 |
5 g | Glass bottle |
578.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96854-34-1 | |
152.19 | |
RRVFYOSEKOTFOG-VIFPVBQESA-N | |
6950537 | |
C1=CC=C(C=C1)C(CCO)O |
C9H12O2 | |
MFCD00221510 | |
1s-1-phenylpropane-1,3-diol, s-1-phenyl-1,3-propanediol, s-1-phenylpropane-1,3-diol, 1s-phenyl-1,3-propanediol, s-1-phenyl-propane-1,3-diol, s, ?-1-phenyl-1,3-propanediol, 1,3-propanediol,1-phenyl-, 1s, 1,3-propanediol, 1-phenyl-, 1s, s---1-phenyl-1,3-propanediol, puriss sum of enantiomers, gc | |
(1S)-1-phenylpropane-1,3-diol |
Specifications
96854-34-1 | |
100.0 | |
White | |
97.5% min. (GC) | |
MFCD00221510 | |
− 54.50 | |
RRVFYOSEKOTFOG-VIFPVBQESA-N | |
(1S)-1-phenylpropane-1,3-diol | |
6950537 | |
98% | |
(S)-1-Phenyl-1, 3-propanediol |
97.5 | |
62.0°C to 66.0°C | |
Authentic | |
C9H12O2 | |
−54.50° (22°C c=1,1,2-dichloroethane) | |
1s-1-phenylpropane-1,3-diol, s-1-phenyl-1,3-propanediol, s-1-phenylpropane-1,3-diol, 1s-phenyl-1,3-propanediol, s-1-phenyl-propane-1,3-diol, s, ?-1-phenyl-1,3-propanediol, 1,3-propanediol,1-phenyl-, 1s, 1,3-propanediol, 1-phenyl-, 1s, s---1-phenyl-1,3-propanediol, puriss sum of enantiomers, gc | |
C1=CC=C(C=C1)C(CCO)O | |
152.19 | |
152.19 | |
Crystalline Powder and/or Chunks |
Safety and Handling
Product Identifier
- (S)-1-Phenyl-1,3-propanediol
RUO – Research Use Only