missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, Thermo Scientific Chemicals Product Code.: 10547410

(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, Thermo Scientific Chemicals

Product Code. 10547410
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10547410

Brand: Thermo Scientific Acros 435830010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104530-79-2
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD08234425,MFCD07778394
InChI Key MIPHRQMEIYLZFZ-UHFFFAOYNA-N
Synonym s-3-aminotetrahydrofuran, s-tetrahydrofuran-3-amine, 3s-oxolan-3-amine, 3-furanamine,tetrahydro-, 3s, s-tetrahydro-furan-3-ylamine, tetrahydrofuran-3beta-amine, s-3-amino-tetrahydrofuran, 5-tetrahydrofuran-3-amine, 3s-tetrahydro-3-furanamine, 3s-tetrahydrofuran-3-amineShow MoreShow Less
PubChem CID 14243168
IUPAC Name oxolan-3-amine
SMILES NC1CCOC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.9°C to 142.8°C
Color White to Brown
Elemental Analysis 48.9 to 53.0 % C 5.2 to 5.6 % N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Specific Rotation -4
Formula Weight 259.32
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material (S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number340411 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number435830010,435830050
Lot Number341180 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number435830010,435830050
compliance-icons
Product Identifier
  • (S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.