missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-4-Boc-(3-hydroxymethyl)morpholine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.
Solubility
Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.
Solubility
Slightly soluble in water.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.

Specifications
Specifications
Melting Point | 80°C (literature) |
Color | White |
Beilstein | 10251196 |
Solubility Information | Slightly soluble in water. |
Quantity | 250 mg |
Formula Weight | 217.27 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | (S)-4-Boc-(3-hydroxymethyl)morpholine |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
81400012 | Certificate of Analysis | 07/12/2021 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(S)-4-Boc-(3-hydroxymethyl)morpholine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction