missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-CLASSIC-90 2-Door Yellow Safety Storage Cabinets
For the safe and approved storage of flammable hazardous substances in working areas, with an optional door open arrest system.
POA
Specifications
Certifications/Compliance | EN 14470-1 |
---|---|
Color | Yellow |
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Triple-hinged |
Product Code | Brand | Load Bearing Capacity | Configuration | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Load Bearing Capacity | Configuration | Includes | Price | Quantity & Availability | |||
15638845
|
Asecos™
3011600233724 |
4 x Drawers 60 kg/29 L each | 4 x 60 kg/29 L sheet steel drawers | Door open arrest system |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
15769428
|
Asecos™
3020500233711 |
6 x Drawers 60 kg/19 L each | 6 x 60 kg/19 L sheet steel drawers | Door open arrest system |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
15879216
|
Asecos™
3011600233719 |
3 x Tray shelves 150 kg/33 L each; Insert 135 kg; Sump 55 L | 3 x 150 kg/33 L sheet steel tray shelves; 1 x 135 kg sheet steel insert; 1 x 55 L sheet steel sump | Door open arrest system |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Aseco’s first EN-compliant safety storage cabinet
- Decades of developing and optimizing ensure that the S-CLASSIC-90 is technically sophisticated
- Available in a range of widths and heights
- Versatile fields of application due to various interior equipment
- Convenient handling – door operation with minimal effort
- Optional door open arrest system allows convenient and safe removal or storage of containers with both hands
- Door lockable with cylinder lock, the cabinets can be integrated by the customer in an existing locking system
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- In accordance with EN 14470-1 and TRGS 510
Specifications
EN 14470-1 | |
Steel | |
Triple-hinged | |
Door open arrest system | |
Powder-coated |
Yellow | |
Safety storage cabinet | |
Storage of flammable and hazardous substances | |
2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Asecos™ S-CLASSIC-90 2-Door Yellow Safety Storage Cabinets