missing translation for 'onlineSavingsMsg'
Learn More
S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals Product Code.: 15461338

S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals

Product Code. 15461338
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15461338

Brand: Thermo Scientific Alfa Aesar H55175.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 934-87-2
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00008752
InChI Key WBISVCLTLBMTDS-UHFFFAOYSA-N
Synonym s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenylShow MoreShow Less
PubChem CID 13630
IUPAC Name S-phenyl ethanethioate
SMILES CC(=O)SC1=CC=CC=C1

Specifications

Color Colorless
Density 1.124 g/mL
Boiling Point 99°C to 100°C (6 mmHg)
Flash Point 79°C (174°F)
Linear Formula CH3COSC6H5
Refractive Index 1.57
Quantity 25 g
Formula Weight 152.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material S-Phenyl thioacetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW16K023 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberH55175.06,H55175.14
Lot NumberM31K009 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog NumberH55175.06,H55175.14
Lot NumberU31J039 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberH55175.06,H55175.14
Lot NumberN17J045 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH55175.14
Lot NumberZ27C033 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55175.06,H55175.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.