Learn More
Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt
Group I agonist
Brand:  Tocris Bioscience™ 0162/10
174.41 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18004662
  • 215.00 € / 10mg
Explore more special offers
Description
Description
S-Sulfo-L-cysteine sodium salt is an EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.
Specifications
Specifications
7381-67-1
Soluble to 100 mM in water
Observed MW: 223.19
10 mg
C3H6NO5S2Na