missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt

Group I agonist

215.00 € - 780.00 €

Specifications

CAS 7381-67-1
Solubility Soluble to 100 mM in water
Molecular Formula C3H6NO5S2Na
Formula Weight Observed MW: 223.19
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18064342
View Documents
Tocris Bioscience™
0162/50
50 mg
780.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
18004662
View Documents
Tocris Bioscience™
0162/10
10 mg
215.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.
Specifications

Specifications

7381-67-1
Soluble to 100 mM in water
C3H6NO5S2Na
Observed MW: 223.19
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at -20°C

SDS
Documents

Documents

Product Certifications

For Research Use Only