missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals S-Trityl-L-cysteine, 97% Product Code.: 15520654

Thermo Scientific Chemicals S-Trityl-L-cysteine, 97%

Product Code. 15520654
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15520654

Brand: Thermo Scientific Chemicals 173010250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C22H21NO2S
MDL Number MFCD00002611

Specifications

Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula (C6H5)3CSCH2CH(NH2)CO2H
Specific Rotation +85° to +89° (24°C, 589nm) (c=1, acetic acid 80%)
Water 0.5% max.
Quantity 25 g
Formula Weight 363.46
Percent Purity ≥96.0%
Physical Form Granular Powder
Chemical Name or Material S-Trityl-L-cysteine, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0437760 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog Number173010050,173010250
Lot NumberA0425870 Certificate TypeCertificate of Analysis Date14/01/2021 Catalog Number173010050,173010250
Lot NumberA0362749 Certificate TypeCertificate of Analysis Date18/08/2020 Catalog Number173010050,173010250
compliance-icons
Product Identifier
  • S-Trityl-L-cysteine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals S-Trityl-L-cysteine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.