Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Component of the system for the safe loosening of stuck ground joints.
Brand:  DWK Life Sciences 286216409
10.60 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15854608
  • 12.20 € / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
  • By means of an even force distribution, stuck joints can be safely loosened.
  • Due to their material properties, pull rings have very good sliding properties.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
To secure the security grinding
Safeguard
NS 14/23