Learn More
DWK Life Sciences Safeguarding, for safety standard ground sockets
Component of the system for the safe loosening of stuck ground joints.
1.31 € - 20.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15854608
|
DWK Life Sciences
286216409 |
NS 14/23 |
12.20 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10151501
|
DWK Life Sciences
286216606 |
NS 29/32 | N/A | ||||||
15981551
|
DWK Life Sciences
286216906 |
NS 19/26 |
12.85 €
Pack of 10 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
|||||
15874608
|
DWK Life Sciences
286217302 |
NS 34/35 |
20.00 €
Pack of 10 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
10161741
|
DWK Life Sciences
286216709 |
NS 45/40 | N/A | ||||||
15901561
|
DWK Life Sciences
286217105 |
NS 24/29 |
14.05 €
Pack of 10 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
Description
Polyamide tension rings have very good sliding properties. They help to safely loosen fixed joints through even distribution of force.
- By means of an even force distribution, stuck joints can be safely loosened.
- Due to their material properties, pull rings have very good sliding properties.
Specifications
10/Pk. | |
Safeguard |