missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Safety Standard Ground Cone, with collar

DURAN™ safety ground joints for increased safety requirements.

Brand:  DWK Life Sciences 286183403

168.30 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15806911

  • 195.00 € / Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ cores with collar are designed for this purpose with an additionally formed glass ring.

  • Core with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
  • Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10/Pk.
With Collar
2 mm
Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.