Learn More
DWK Life Sciences Safety Standard Ground Cone, with collar
DURAN™ safety ground joints for increased safety requirements.
96.70 € - 281.00 €
Specifications
Quantity 10/Pk.
Type With Collar
View More Specs
Products 6
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15806911
View Documents
DWK Life Sciences
286183403
32 mm
195.00 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15816911
View Documents
DWK Life Sciences
286184502
40 mm
281.00 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15866901
View Documents
DWK Life Sciences
286181402
13 mm
96.70 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15876901
View Documents
DWK Life Sciences
286181908
17 mm
108.00 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15886901
View Documents
DWK Life Sciences
286182407
22 mm
104.00 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
15896901
View Documents
DWK Life Sciences
286182904
26 mm
112.00 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
Description
Description
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ cores with collar are designed for this purpose with an additionally formed glass ring.
  • Core with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
  • Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
  • Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
10/Pk.
With Collar