Learn More
DWK Life Sciences Safety Standard Ground Cone, with collar
DURAN™ safety ground joints for increased safety requirements.
Brand: DWK Life Sciences 286182904
Description
Securing joints by using our KECK™-products is a well-known and proven method. In order to meet high safety demands, e.g. when dealing with hazardous substances or pressure differences, DURAN™ Safety Joints offer very good protection against accidental loosening of the connection. Furthermore, DURAN™ Safety Joints offer a system to separate jammed cones and sockets. This minimizes the risk of breakage and related threats. The DURAN™ cores with collar are designed for this purpose with an additionally formed glass ring.
- Core with collar and additionally moulded glass ring prevents unintentional loosening of the ground joint
- Firmly seated ground joints can be loosened more easily here. The danger of glass breakage and associated risks is thus minimized.
- Use in equipment for handling hazardous substances or working with pressure differences.g in the high vacuum range
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
10/Pk. | |
With Collar | |
2 mm |
Use in equipment for handling hazardous substances or working with pressure differences in the high vacuum range; When working under increased safety requirements; Installation in glass apparatus and devices | |
26 mm |