Learn More
SCAT HPLC Safety Waste Caps, V2.0
GREENER_CHOICE
SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.
106.00 € - 210.00 €
Specifications
Type Safety Waste Cap
For Use With (Application) Waste Disposal System
For Use With (Equipment) Waste Disposal Container
View More Specs
Products 10
Product Code Brand Thread Size Includes Price Quantity & Availability  
Product Code Brand Thread Size Includes Price Quantity & Availability  
17122576
GREENER_CHOICE
View Documents
SCAT
307108
S40/GL40 3 x 2.3/3.2 mm OD + connector for exhaust filter
128.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
16443809
GREENER_CHOICE
View Documents
SCAT
307923
GL 45 2 x PFA-fitting 2.3/3.2 mm OD, 2 x PFA-blind plug, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter
109.00 €
Each
In Stock
to see stock.
Edge
Add to basket
16413809
GREENER_CHOICE
View Documents
SCAT
307912
GL 45 3 x PFA-fitting 2.3/3.2 mm OD, 1 x PFA-blind plug + connector for exhaust filter
106.00 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
16439963
GREENER_CHOICE
View Documents
SCAT
308921
GL 45 4 x PFA-fitting 1.6/2.3/3.2 mm OD, 4 x PFA-blind plug, 1 x 5.0 - 11.5 mm ID, 1 x PTFE-blind plug + connector for exhaust filter
161.00 €
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
16526034
GREENER_CHOICE
View Documents
SCAT
307925
S60/61 2 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter
148.00 €
Each
In Stock
to see stock.
Edge
Add to basket
16635722
GREENER_CHOICE
View Documents
SCAT
307500
S60 3 x 1.6/2.3/3.2 mm OD, 3 x PFA-blind plug, 3 x 5.0 - 11.5 mm ID, 3 x PTFE-blind plug + connector for exhaust filter
194.00 €
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
16626214
GREENER_CHOICE
View Documents
SCAT
307931
S60/61 4 x 2.3/3.2 mm OD, 1 x 5.0 - 11.5 mm ID + connector for exhaust filter
183.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
17089693
GREENER_CHOICE
View Documents
SCAT
307918
S60/61 3 x 2.3/3.2 mm OD + connector for exhaust filter
152.00 €
Each
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
17175501
GREENER_CHOICE
View Documents
SCAT
307916
S60/61 3 x 2.3/3.2 mm OD, grounding connection + connector for exhaust filter
210.00 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
18696468
View Documents
SCAT
307970
S60/61 1x NPT 3/8 in. (f), 1x connection for exhaust filter (45° beveled)
196.00 €
1 sheet
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
Description
Description
  • Discover the flexibility of the world's leading system for solvent safety
  • SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
  • Optimum protection when collecting hazardous or toxic liquid waste
  • Flexible system with fittings and tube connectors in different sizes
  • With multiple options to connect different tubing and capillary sizes
  • Time saving in changing the waste containers while tubes are fixed
  • Resistant against aggressive media
  • PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
  • Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
  • PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
  • Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
  • Flammability classification V-0 (UL-94)
  • Insertion Label: Mark your bottles with individual descriptions
  • PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
  • Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
  • Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications
Specifications
Safety Waste Cap
Waste Disposal System
Waste Disposal Container
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
SCAT HPLC Safety Waste Caps, V2.0