missing translation for 'onlineSavingsMsg'
Learn More

SCAT HPLC Safety Waste Caps, V2.0

SCAT Safety Waste Caps protect staff and the environment from harmful solvent vapours. With a suitable exhaust filter, they turn your disposal container into a closed system without escaping solvent vapours.

Brand:  SCAT 307970

 View more versions of this product

Product Code. 18696468

  • 196.00 € / 1 sheet
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Discover the flexibility of the world's leading system for solvent safety
  • SafetyWasteCap with S 60/61 thread in the new design of generation 2.0
  • Optimum protection when collecting hazardous or toxic liquid waste
  • Flexible system with fittings and tube connectors in different sizes
  • With multiple options to connect different tubing and capillary sizes
  • Time saving in changing the waste containers while tubes are fixed
  • Resistant against aggressive media
  • PTFE Body 360° freely rotatable: Provides easy container change without distortion of tubing
  • Body with integrated FKM Seal: Provides hermetical connection to the container opening and adapts smoothly even to rugged glass surfaces. An additional PTFE layer protects the seal from direct contact to solvents.
  • PFA Fittings: Improved design for easy tubing connection - Excellent chemical resistance - Flammability classification V-0 (UL-94)
  • Medical Grade PTFE: Excellent chemical resistance Food safe FDA compliant -
  • Flammability classification V-0 (UL-94)
  • Insertion Label: Mark your bottles with individual descriptions
  • PPS Screw Cap: Autoclavable / sterilisable up to 200°C - Flammability classification V-0 (UL-94)
  • Ergonomic Design: Enhanced stability and construction - Improved handling - Easy installation - Smart exchange
  • Huge range of accessories, optional: exhaust filters, funnel use, level control preventing overflowing containers
Specifications

Specifications

Waste Disposal System
1x NPT 3/8 in. (f), 1x connection for exhaust filter (45° beveled)
S60/61
Waste Disposal Container
PTFE-EL
Safety Waste Cap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCAT HPLC Safety Waste Caps, V2.0 > Thread Size: S60/61, Includes: 1x NPT 3/8 in. (f) , 1x connection for exhaust filter (45° beveled)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.