missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SafTest™ Dispenser Set
Dispenser Set is an accessory for SafTest™ test kits.
1915.00 € - 3405.00 €
Specifications
Storage Requirements | Room temperature |
---|
Includes: One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | ||||||
15732422
|
MP Biomedicals™
07DSAK2000 |
Alkenal Testing |
2815.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15742422
|
MP Biomedicals™
07DSFA2000 |
Free Fatty Acid Testing |
2820.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15772422
|
MP Biomedicals™
07DSPR2000 |
Peroxide Testing |
3405.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15762422
|
MP Biomedicals™
07DSPF2000 |
Percent Fat Testing |
2160.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15782422
|
MP Biomedicals™
07DSTG2000 |
Total Glycerin Testing |
1915.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
15752422
|
MP Biomedicals™
07DSMA2000 |
Malonaldehyde Testing |
2650.00 €
1 set |
Estimated Shipment: 12-06-2025
Log in to see stock. |
||||||
Description
- Dispenser set for each chosen endpoint
- Allows for accurate testing, every time
- For use with SafTest™ kits
Specifications
Room temperature |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MP Biomedicals™ SafTest™ Dispenser Set