missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SafTest™ Dispenser Set

Dispenser Set is an accessory for SafTest™ test kits.

Brand:  MP Biomedicals™ 07DSPR2000

 View more versions of this product

Product Code. 15772422

  • 3405.00 € / 1 set
Estimated Shipment: 17-06-2025
to see stock.
Explore more special offers
Add to basket

Includes: One dispenser for each reagent (A, B, and preparation Reagent) and one reagent holding rack
This item is not returnable. View return policy

Description

Description

  • Dispenser set for each chosen endpoint
  • Allows for accurate testing, every time
  • For use with SafTest™ kits
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Peroxide Testing
Room temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ SafTest™ Dispenser Set > SafTest™ Dispenser Set; For use with: Peroxide Test Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.