missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SAHARA PPO S14P Heated Bath Circulators

Ideal when visibility of your application is required. These baths maintain temperatures from ambient +13° to 100°C, with maximum bath volume of 14 liters.

Brand:  Thermo Scientific™ 1523141

 View more versions of this product

Product Code. 11318350

  • 1819.05 € / Each
Estimated Shipment: 20-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

SC100
14 L
USB
Force Pump, Suction Pump
17.8 x 14.1 x 14.2 in. (45.2 x 35.8 x 36.1 cm)
45.2 cm
35.8 cm
36.1 cm
6.3 kg
230 V
SC100-S14P; 230V/50Hz
CE/ROHS/WEEE
17 L/min.
Ambient +13° to 100°C
17.8 in.
14.1 in.
14.2 in.
13.9 lb.
230 V 50 Hz
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SAHARA PPO S14P Heated Bath Circulators >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.