Learn More
Thermo Scientific™ Precision™ Circulating Water Baths
Achieve outstanding temperature uniformity for your samples with perimeter-directed water flow on these circulating water baths. Featuring advanced temperature controller.
Brand: Thermo Scientific™ TSCIR19
2242.12 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
Ensure temperature uniformity and control with theseThermo Scientific™ Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
- Achieve enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Optimize scheduling with auto-on and auto-off timers
- Accommodate taller labware with new hinged lid and extended height
- Help prevent bath overheating and damage with low-fluid protection
- Easily operate and monitor with icon-based graphical display
- Protect your work with audible alarms

Specifications
5.0 gal. | |
19L Circulating Water Bath | |
UL, CE | |
Ambient +5° to 100°C | |
1200 w | |
304 Stainless Steel | |
4 | |
±0.05°C | |
15.5 x 24.9 x 9.8 in. (394 x 632 x 249 mm) | |
9.0-10.5 / 4.3-5.0 (120V/230V) |
Digital | |
19 L | |
Digital | |
Monochrome LCD | |
1 Stainless Steel Gable cover and Rubber Duck | |
Precision CIR 19 | |
±0.1°C | |
12 x 15.3 x 7.6 in. (305 x 387 x 192 mm) | |
Global Voltage 100/115 V 200/230 V 50/60 Hz |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.