Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
Brand:  Saint-Gobain 96430-16
 View more versions of this product
Product Code. 11740418
  • 930.00 € / 15m
Estimated Shipment: 07-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
  • Does not contaminate fluids
  • Superior flex life in peristaltic pumps
  • Chemically resistant to a wide range of fluids
  • Crystal clear for easy visual flow monitoring
  • Meets FDA criteria for food contact
  • Will not degrade over time
  • Will not release hazardous byproducts when properly incinerated
  • Does not yield any taste
  • Helps ensure accurate results from analytical instrument tests
†At 73°F (23°C).
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
Specifications
Specifications
Compliant with FDA 21 CFR, 177.2600
19.1 mm
25.4 mm
1/8 in.
3/4 in.
1 in.
50 ft.
3.2 mm