missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
94.48 € - 930.00 €
Specifications
Certifications/Compliance | Compliant with FDA 21 CFR, 177.2600 |
---|---|
Overall Length (English) | 50 ft. |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
11780408
|
Saint-Gobain
MFLX96430-04 (EA) |
3/16 in. | 5/16 in. | 1/16 in. | |||||
11700418
|
Saint-Gobain
MFLX96430-08 (EA) |
5/16 in. | 7/16 in. | 1/16 in. | |||||
11790408
|
Saint-Gobain
MFLX96430-06 (EA) |
1/4 in. | 3/8 in. | 1/16 in. | |||||
11760408
|
Saint-Gobain
MFLX96430-00 (EA) |
1/16 in. | 3/16 in. | 1/16 in. | |||||
11770408
|
Saint-Gobain
MFLX96430-02 (EA) |
1/8 in. | 1/4 in. | 1/16 in. | |||||
11710418
|
Saint-Gobain
MFLX96430-10 (EA) |
3/8 in. | 1/2 in. | 1/16 in. | |||||
11720418
|
Saint-Gobain
MFLX96430-12 (EA) |
1/2 in. | 3/4 in. | 1/8 in. | |||||
11730418
|
Saint-Gobain
96430-14 |
5/8 in. | 7/8 in. | 1/8 in. | |||||
11740418
|
Saint-Gobain
96430-16 |
3/4 in. | 1 in. | 1/8 in. |
930.00 €
15m |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
- Does not contaminate fluids
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
- Crystal clear for easy visual flow monitoring
- Meets FDA criteria for food contact
- Will not degrade over time
- Will not release hazardous byproducts when properly incinerated
- Does not yield any taste
- Helps ensure accurate results from analytical instrument tests
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
Specifications
Compliant with FDA 21 CFR, 177.2600 | |
50 ft. |