missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ 2375 Ultra Chemical Resistant Tubing

Tygon™ 2375 Ultra Chemical Resistant Tubing for Chemical Transfer Applications.

Brand:  Saint Gobain Life Sciences 95630-11

 View more versions of this product

Product Code. 11944438

  • 132.00 € / 15m
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.

Tygon™ 2375 tubing from Saint-Gobain is ideal for applications where harsh cleaning chemicals are used. It is clear, flexible, and virtually unaffected by acids, bases, ketones, salts, and alcohols. This product is entirely free of plasticizers and eliminates fluid contamination, premature brittleness and cracking that often occurs with other flexible tubing products. When properly incinerated, Tygon™ 2375 does not release any pollution that could harm the environment.

  • Outstanding chemical resistance
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Safer disposal – releases no harmful and corrosive hydrogen chloride gas
  • Smoother inner surface – provides better flow and inhibits particulate buildup
  • Low sorption – minimizes cross-contamination and maintains media purity
  • Clear tubing for easier and better observation

Typical Applications

  • Industrial/institutional cleaning
  • Chemical transfer systems
  • Transfer lines for chemical analyzers
  • Detergent supply lines
  • Ink and printing fluid dispensing
  • Industrial X-ray processing
  • Power generation sampling and drain lines
  • Paint and solvent production
  • Hazardous materials handling
Specifications

Specifications

Transfer Tubing
1.6 mm
3/16 in.
50 ft.
0.0625 in.
Clear
0.063 in.
Tygon
4.8 mm
15 m
1.6 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.