missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ 2375 Ultra Chemical Resistant Tubing

Tygon™ 2375 Ultra Chemical Resistant Tubing for Chemical Transfer Applications.

132.00 € - 780.00 €

Specifications

Product Type Transfer Tubing
Material Tygon
Color Clear
View More Specs

Products 6
Product Code Brand Diameter (English) Inner Diameter (English) Outer Thickness (Metric) Thickness (English) Price Quantity & Availability  
Product Code Brand Diameter (English) Inner Diameter (English) Outer Thickness (Metric) Thickness (English) Price Quantity & Availability  
11944438
View Documents
Saint Gobain Life Sciences
95630-11
0.063 in. 3/16 in. 1.6 mm 0.0625 in.
132.00 €
15m
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
11994438
View Documents
Saint Gobain Life Sciences
95630-16
1/4 in. 0.375 in. 1.6 mm 0.0625 in.
235.00 €
15m
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
11984438
View Documents
Saint Gobain Life Sciences
95630-15
0.188 in. 5/16 in. 1.6 mm 0.0625 in.
237.00 €
15m
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
11904448
View Documents
Saint Gobain Life Sciences
AJK00022
5/16 in. 7/16 in. 1.6 mm 0.0625 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11934448
View Documents
Saint Gobain Life Sciences
AJK00038
0.5 in. 0.75 in. 3.2 mm 1/8 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11944448
View Documents
Saint Gobain Life Sciences
95630-21
0.625 in. 0.875 in. 3.2 mm 1/8 in.
780.00 €
15m
Estimated Shipment: 07-04-2025
to see stock.
Add to basket
 
Description

Description

Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.

Tygon™ 2375 tubing from Saint-Gobain is ideal for applications where harsh cleaning chemicals are used. It is clear, flexible, and virtually unaffected by acids, bases, ketones, salts, and alcohols. This product is entirely free of plasticizers and eliminates fluid contamination, premature brittleness and cracking that often occurs with other flexible tubing products. When properly incinerated, Tygon™ 2375 does not release any pollution that could harm the environment.

  • Outstanding chemical resistance
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Safer disposal – releases no harmful and corrosive hydrogen chloride gas
  • Smoother inner surface – provides better flow and inhibits particulate buildup
  • Low sorption – minimizes cross-contamination and maintains media purity
  • Clear tubing for easier and better observation

Typical Applications

  • Industrial/institutional cleaning
  • Chemical transfer systems
  • Transfer lines for chemical analyzers
  • Detergent supply lines
  • Ink and printing fluid dispensing
  • Industrial X-ray processing
  • Power generation sampling and drain lines
  • Paint and solvent production
  • Hazardous materials handling
Specifications

Specifications

Transfer Tubing
Tygon
Clear
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Saint Gobain Life Sciences Tygon™ 2375 Ultra Chemical Resistant Tubing

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.