missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%, Thermo Scientific Chemicals

16.73 € - 745.71 €

Chemical Identifiers

CAS 90-02-8, 108-95-2
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10745791
View Documents
Thermo Scientific Acros
132600020
2 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10071290
View Documents
Thermo Scientific Acros
132601000
100 g Glass bottle
34.97 €
100g
Please to purchase this item. Need a web account? Register with us today!
10367160
View Documents
Thermo Scientific Acros
132602500
250 g Glass bottle
60.80 €
250g
Please to purchase this item. Need a web account? Register with us today!
10234900
View Documents
Thermo Scientific Acros
132600010
1 kg Glass bottle
191.96 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11944791
View Documents
Thermo Scientific Acros
132600050
5 kg Plastic drum
745.71 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90-02-8, 108-95-2
122.12
SMQUZDBALVYZAC-UHFFFAOYSA-N
6998
2-hydroxybenzaldehyde
C7H6O2
MFCD00003317
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
CHEBI:16008
OC1=CC=CC=C1C=O

Specifications

90-02-8, 108-95-2
1.1600g/mL
99%
2-(HO)C6H4CHO
MFCD00003317
salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
OC1=CC=CC=C1C=O
122.12
CHEBI:16008
≥98.5%
Salicylaldehyde
Yellow
Authentic
C7H6O2
1.5720 to 1.5740 (20°C, 589nm)
1.16 to 1.17
SMQUZDBALVYZAC-UHFFFAOYSA-N
2-hydroxybenzaldehyde
6998
122.12
Liquid
Videos
Safety and Handling

Safety and Handling

Danger

Recommended Storage : product darkens during storage

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.