Learn More
Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents
Brand: Fisher BioReagents BP1325-1
This item is not returnable.
View return policy
Description
Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.Specifications
Saline-Sodium Citrate | |
Liquid | |
7 | |
1L |
RT | |
DNase free | |
Molecular Biology | |
2.9 to 3.1M |
Safety and Handling
Product Identifier
- Saline-Sodium Citrate 20X Solution