missing translation for 'onlineSavingsMsg'
Learn More

Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents

43.85 € - 195.00 €

Specifications

Chemical Name or Material Saline-Sodium Citrate
Content And Storage RT
Form Liquid
DNase DNase free
pH 7
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10785104
View Documents
Fisher BioReagents
BP1325-20
20L
195.00 €
20 L
Please to purchase this item. Need a web account? Register with us today!
10418353
View Documents
Fisher BioReagents
BP1325-1
1L
43.85 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.
Specifications

Specifications

Saline-Sodium Citrate
Liquid
7
2.9 to 3.1M
RT
DNase free
Molecular Biology
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Saline-Sodium Citrate 20X Solution

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.