Learn More
Thermo Scientific™ Sample Racks for Precision™ Water Baths
Keep your samples in place with racks for water baths.
53.16 € - 462.30 €
Products 14
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15395897
View Documents
Thermo Scientific™
3161601
Stainless Steel Test Tube Rack
462.30 €
Each
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
15385897
View Documents
Thermo Scientific™
3166183
Stainless Steel Petri Dish Rack
362.81 €
Each
Estimated Shipment: 21-08-2025
to see stock.
Add to basket
15305917
View Documents
Thermo Scientific™
3166184
0.5mL Microfuge Rack (Requires Clip Fastener)
278.39 €
Each
Estimated Shipment: 07-08-2025
to see stock.
Add to basket
15315917
missing translation for 'viewDocuments'
Thermo Scientific™
3166185
1.5mL Microfuge Rack (Requires Clip Fastener)
264.32 €
Each
Estimated Shipment: 07-08-2025
to see stock.
Add to basket
15355907
missing translation for 'viewDocuments'
Thermo Scientific™
5970-0513TC
Nalgene Unwire Full Test tube Rack - Red 72x13mm
73.67 €
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15365907
missing translation for 'viewDocuments'
Thermo Scientific™
5970-0516TC
Nalgene Unwire Full Test tube Rack - Red 72x16mm
72.66 €
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15375907
missing translation for 'viewDocuments'
Thermo Scientific™
5970-0520TC
Nalgene Unwire Full Test tube Rack - Red 40x20mm
73.67 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15385907
View Documents
Thermo Scientific™
5970-0525TC
Nalgene Unwire Full Test tube Rack - Red 40x25mm
76.88 €
Each
Estimated Shipment: 18-07-2025
to see stock.
Add to basket
15395907
View Documents
Thermo Scientific™
5970-0530TC
Nalgene Unwire Full Test tube Rack - Red 24x30mm
79.70 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15305907
View Documents
Thermo Scientific™
5972-0013TC
Nalgene Half Test tube Rack - White 36x13mm
53.16 €
Each
Estimated Shipment: 04-07-2025
to see stock.
Add to basket
15315907
View Documents
Thermo Scientific™
5972-0016TC
Nalgene Half Test tube Rack - White 36x16mm
55.78 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15325907
View Documents
Thermo Scientific™
5972-0020TC
Nalgene Half Test tube Rack - White 20x20mm
55.17 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15345907
View Documents
Thermo Scientific™
5972-0030TC
Nalgene Half Test tube Rack - White 9x30mm
73.57 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
15335907
View Documents
Thermo Scientific™
5972-0025TC
Nalgene Half Test tube Rack - White 16x25mm
64.72 €
Each
Estimated Shipment: 09-07-2025
to see stock.
Add to basket
Description
Description
Keep your samples in place with racks for Thermo Scientific™ Precision™ water baths.
Stainless-Steel Petri Dish Rack
  • Measures 9.75 x 8.75 x 6.25 in. (24.8 x 22.2 x 15.9cm)
  • Holds 80 x 50mm or 30 x 90mm Petri dishes
Stainless-Steel Test Tube Rack
  • Measures 10 x 8.5 x 5.5 in. (25.4 x 21.6 x 14cm)
  • Openings are 1in. (2.5cm) square
  • Holds 50 test tubes up to 25mm from 97 to 150mm high
Microcentrifuge Test Tube Racks
  • Requires Clip Fastener
  • Measures 127 x 101 x 39mm (5 x 4 x 1.5 in.)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Sample Racks for Precision™ Water Baths