Learn More
Masterflex™ Sterilized Sampling Bottle Assemblies Product Code.: 13286950
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
Masterflex™ Sterilized Sampling Bottle Assemblies
Product Code. 13286950
500 mL
Click to view available options
Capacity (Metric):
500 mL
Product Code. 13286950
Brand: Masterflex™ 9564404
Estimated Shipment: 26-05-2025
to see stock.
Add to basket
Add to basket
Transfer, sample and store biopharmaceutical products and reagents in applications from research to process with these sample bottles. Cole-Parmer™ Sterilized Sampling Bottle Assemblies are pre-sterilized by gamma irradiation.
Secure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connections.
Specifications
Capacity (Metric) 500 mL
For Use With (Application) Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to process.
Material Plastic
Closure Included Yes
Closure Liner Seamless
Product Content Correction
Product Title
Masterflex™ Sterilized Sampling Bottle Assemblies >