missing translation for 'onlineSavingsMsg'
Learn More

Notrax Sani-Master™ Disinfecting Foot Bath with Drying Entrance Mat, Charcoal/Black

This Notrax Sani-Master™ 19-mm thick extendable mat cleans, sanitizes and dries shoes, recommended at entrances to prevent contamination. As traffic passes, rubber tips bend to immerse soles in disinfectant solution, absorbing mat section cleans and dries shoes to prevent slips.

Brand:  Notrax 348S0033CH

 View more versions of this product

Product Code. 17702028

  • 164.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Combining three of Notrax™ matting features of sanitizing, absorbing and modularity into a complete disinfection entrance mat solution, Sani-Master™ is a 19 mm thick extendable mat that cleans, sanitizes and dries shoes
  • Traditionally used in the food industry, Sani-Master™ has become mainstream as part of general hygiene protocols and is recommended at entrances of schools, hotels, gyms, and offices, as well as pharmacies, health care facilities, and hospitals to prevent contamination from entering the building
  • Section 1 incorporates the Sani-Trax™ disinfection foot bath with flexible bristles that bend to immerse soles in disinfectant solution to wash and disinfect shoes as traffic passes
  • Section 2 is an extendable drying zone to dry shoes and prevent slips with patented connection system that is seamless to avoid changes in height or disconnection of the mats
  • Section 3 is an end drying zone with integrated bevelled edges to prevent tripping
  • The drying carpets can be fully customized with a logo, design or used for communication of hygiene protocols such as hand disinfection and social distancing
Specifications

Specifications

Charcoal/Black
105 cm
Industrial and Commercial Matting
3.4 ft.
Natural Rubber
91 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.