Learn More
Thermo Scientific Chemicals Sapphire substrate, 10x10x0.432mm, EPI polished two sides, C-axis, LED grade
Brand:  Thermo Scientific Chemicals 045017.KU
617.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15426675
  • 620.09 € / Pack of 5
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sapphire Substrate
EPI polished two sides
5 Ea.
10 x 10 x 0.432 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD25S048 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045018.KT,045017.KU,045017.KT
Lot NumberG03S019 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045018.KT,045017.KU,045017.KT
Lot NumberQ20H037 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045018.KT,045017.KU,045017.KT
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific Chemicals Sapphire substrate, 10x10x0.432mm, EPI polished two sides, C-axis, LED grade > 5each