Learn More
Sartorius Absorbent Pads
presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device
Brand: Sartorius 15410--47----ALR
This item is not returnable.
View return policy
Description
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
- 47mm has approx. 3ml absorption capacity
- 50mm with approx. 3.5ml absorption capacity
Specifications
Nutrient Pad | |
Packaged in 10 magazines | |
1.85 in. | |
4.7 cm |