Learn More
Sartorius Absorbent Pads
presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device
110.00 € - 288.00 €
Product Code | Brand | Packaging Type | Diameter (English) Outside | Diameter (Metric) Outside | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Diameter (English) Outside | Diameter (Metric) Outside | Price | Quantity & Availability | |||
11780686
|
Sartorius
15400--50------N |
Packaged in petridish | 1.96 in. | 5 cm | N/A | ||||
11730876
|
Sartorius
15410--47----ALR |
Packaged in 10 magazines | 1.85 in. | 4.7 cm | N/A | ||||
11740876
|
Sartorius
15410--50----ALR |
Packaged in 10 magazines | 1.96 in. | 50 mm | N/A | ||||
Description
Sartorius 1.4mm thick absorbent pads are wetted with the appropriate liquid culture medium before a membrane filter is placed on them. They come presterilized in plastic magazines, which fit onto the Sartorius manual dispensing device.
The absorbent pads are available in two diameters:
- 47mm has approx. 3ml absorption capacity
- 50mm with approx. 3.5ml absorption capacity
Specifications
Nutrient Pad |