Learn More
Sartorius Accessories for Biosart™ 100 Membrane Filtration
Biosart™ 100 Membrane Filtration is designed specifically for microbiological testing of pharmaceuticals, food, beverages, water and other liquids.
Brand: Sartorius 16835
154.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Accessories Descriptions
Adapter, ABS copolymer - 14-555-780- Biosart 100 Membrane Lifter
- For easy transfer of the membrane onto agar
- Biosart 100 Adapter
- Fits Biosart 100 Monitor onto 50 mm supports
- Biosart 100 Adapter
- 50 mm stainless-steel frit and silicone gasket
- Fits Biosart 100 Monitor onto 16840 or 16841 base
- Biosart 100 Adapter to accommodate various funnel types
- Fits Biosart 100 Monitors onto Combisart™ manifolds 16844, 16842 and 16843
- Replacement stopper for 16385
- 16385 adapts Biosart 100 Monitors on Combisart manifold

Specifications
Reusable | |
Sartorius Biosart 100 | |
Non-sterile | |
Adapter |
Manifold | |
Stainless Steel | |
Fits Biosart 100 Monitors onto Combisart manifolds 16844, 16842 and 16843 |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.